Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017016494.1 | XP_016871983.1 | 1 | 73 | 101 | |
| GenBank mRNA | AY505173.1 | - | 1 | 45 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylinositol 4-kinase type 2 alpha | PI4K2A |
PI4KII, PIK42A |
-10.7kb | Anti-sense | Chr. 10 - 97640686 - 97676434 on NCBI Build GRCh38 |
| Downstream | MARVEL domain containing 1 | MARVELD1 |
GB14, MARVD1, MRVLDC1, bA548K23.8 |
26.3kb | Anti-sense | Chr. 10 - 97713708 - 97718152 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017016494.1 transcripts.
Set Membership: |
|

AVPI1
Genomic Map