Assay Details
| Gene Symbol: LOC107984638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017021842.1 | XP_016877331.1 | 2 | 1085 | 101 | |
| GenBank mRNA | BC000005.1 | - | 1 | 952 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1629 | LINC01629 | linc-KIAA1737-2 | -58.1kb | Sense | Chr. 14 - 76959638 - 76965802 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105370579 | LOC105370579 | 3.3kb | Anti-sense | Chr. 14 - 77032368 - 77035652 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC107984638
Genomic Map