Assay Details
| Gene Symbol: PARTICL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038942.1 | 1 | 1153 | 75 | ||
| GenBank mRNA | BC009563.1 | - | 1 | 1153 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH2 domain containing 6 | SH2D6 | -100.4kb | Anti-sense | Chr. 2 - 85418931 - 85437029 on NCBI Build GRCh38 | |
| Downstream | methionine adenosyltransferase 2A | MAT2A |
MATA2, MATII, SAMS2 |
92.0 | Anti-sense | Chr. 2 - 85538978 - 85545280 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002042.hg.1
Set Membership: |
|

PARTICL
Genomic Map