Assay Details
| Gene Symbol: NAV2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC018444.1 | - | 1 - 2 | 113 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuron navigator 2 | NAV2 |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Sense | Chr. 11 - 19345200 - 20121601 on NCBI Build GRCh38 | |
| Downstream | NAV2 antisense RNA 5 | NAV2-AS5 | 117.7kb | Anti-sense | Chr. 11 - 19502672 - 19507229 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

NAV2-IT1
Genomic Map