Assay Details
| Gene Symbol: LINC01613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132393.1 | 1 | 2642 | 69 | ||
| GenBank mRNA | BC033399.1 | - | 1 | 2645 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BMP/retinoic acid inducible neural specific 1 | BRINP1 |
DBC1, DBCCR1, FAM5A |
-565.6kb | Anti-sense | Chr. 9 - 119166630 - 119369461 on NCBI Build GRCh38 |
| Downstream | microRNA 147a | MIR147A |
MIR147, MIRN147, hsa-mir-147a |
307.1kb | Anti-sense | Chr. 9 - 120244979 - 120245050 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01613
Genomic Map