Assay Details
| Gene Symbol: LOC102724659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126000.1 | 1 - 2 | 493 | 92 | ||
| GenBank mRNA | BC034298.1 | - | 1 - 2 | 493 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | angiotensin II receptor associated protein | AGTRAP | ATRAP | -27.9kb | Anti-sense | Chr. 1 - 11736085 - 11750771 on NCBI Build GRCh38 |
| Downstream | methylenetetrahydrofolate reductase (NAD(P)H) | MTHFR | 6.1kb | Sense | Chr. 1 - 11785730 - 11806103 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724659
Genomic Map