Assay Details
| Gene Symbol: LOC101928730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120422.1 | 2 - 3 | 861 | 102 | ||
| GenBank mRNA | BC034786.1 | - | 2 - 3 | 861 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1070 | LINC01070 | -70.1kb | Anti-sense | Chr. 13 - 112197333 - 112201000 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1043 | LINC01043 | 41.5kb | Anti-sense | Chr. 13 - 112313467 - 112321766 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928730
Genomic Map