Assay Details
| Gene Symbol: LOC105377938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035649.1 | - | 1 | 655 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 22 member 16 | SLC22A16 |
CT2, FLIPT2, HEL-S-18, OAT6, OCT6, OKB1, dJ261K5.1 |
-1.3kb | Anti-sense | Chr. 6 - 110424587 - 110476641 on NCBI Build GRCh38 |
| Downstream | cyclin dependent kinase 19 | CDK19 |
CDC2L6, CDK11, bA346C16.3 |
130.5kb | Anti-sense | Chr. 6 - 110609978 - 110816531 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105377938
Genomic Map