Assay Details
| Gene Symbol: ZMAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022470.3 | NP_071915.1 | 6 | 7277 | 94 | |
| NM_152240.2 | NP_689426.1 | 7 | 7395 | 94 | ||
| XM_005247706.2 | XP_005247763.1 | 6 | 7685 | 94 | ||
| XM_011513073.1 | XP_011511375.1 | 7 | 7809 | 94 | ||
| GenBank mRNA | BC036485.1 | - | 1 | 1152 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNMB2 antisense RNA 1 | KCNMB2-AS1 | -156.8kb | Sense | Chr. 3 - 178525467 - 178860405 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928739 | LOC101928739 | 81.3kb | Sense | Chr. 3 - 179101371 - 179147973 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZMAT3
Genomic Map