Assay Details
| Gene Symbol: LINC01361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110632.1 | 1 - 2 | 62 | 102 | ||
| GenBank mRNA | BC037222.1 | - | 1 - 2 | 62 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1362 | LINC01362 | Anti-sense | Chr. 1 - 82903183 - 82997971 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC101927587 | LOC101927587 | 590.9kb | Sense | Chr. 1 - 83575787 - 83860996 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209633.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209637.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209635.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209636.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209638.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209634.hg.1 | TC0100014735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000421931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | ENST00000452834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110632.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014735.hg.1 | NR_110633.1 | Refseq |

LINC01361
Genomic Map