Assay Details
| Gene Symbol: LINC01153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037739.1 | - | 1 - 2 | 279 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 11 | WDR11 |
BRWD2, DR11, HH14, SRI1, WDR15 |
-269.2kb | Sense | Chr. 10 - 120851175 - 120909526 on NCBI Build GRCh38 |
| Downstream | fibroblast growth factor receptor 2 | FGFR2 |
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25 |
292.4kb | Anti-sense | Chr. 10 - 121478330 - 121598656 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01153
Genomic Map