| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000462186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000462186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000462186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000462186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000471224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000471224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000471224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000471224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000471224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000472297.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000472297.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000472297.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000472297.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000475499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000475499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000484897.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
ENST00000484897.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011698.hg.1 |
NR_003662.3 |
Refseq |