Assay Details
| Gene Symbol: LOC101927053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110209.1 | 1 - 2 | 334 | 78 | ||
| GenBank mRNA | BC068584.1 | - | 1 - 2 | 334 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | semaphorin 4C | SEMA4C |
M-SEMA-F, SEMACL1, SEMAF, SEMAI |
-48.2kb | Anti-sense | Chr. 2 - 96859736 - 96870943 on NCBI Build GRCh38 |
| Downstream |
glycerol-3-phosphate acyltransferase 2, mitochondrial pseudogene |
LOC653924 | 161.5kb | Sense | Chr. 2 - 97081436 - 97083472 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927053
Genomic Map