Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR4290 host gene | MIR4290HG | -116.5kb | Sense | Chr. 9 - 90020654 - 90041499 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1508 | LINC01508 | 142.3kb | Sense | Chr. 9 - 90300896 - 90433489 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map