Assay Details
| Gene Symbol: LOC105372457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057976.1 | - | 1 | 1051 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NLR family pyrin domain containing 12 | NLRP12 |
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2 |
-29.2kb | Sense | Chr. 19 - 53793584 - 53824403 on NCBI Build GRCh38 |
| Downstream | myeloid associated differentiation marker | MYADM | SB135 | 10.6kb | Anti-sense | Chr. 19 - 53865584 - 53876435 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105372457
Genomic Map