Assay Details
| Gene Symbol: UROS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090736.1 | - | 1 | 1052 | 58 | |
| AK092076.1 | - | 1 | 1430 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | matrix metallopeptidase 21 | MMP21 |
HTX7, MMP-21 |
-3.4kb | Sense | Chr. 10 - 125766453 - 125775821 on NCBI Build GRCh38 |
| Downstream | microRNA 4484 | MIR4484 | mir-4484 | 29.0kb | Anti-sense | Chr. 10 - 125819740 - 125819822 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

UROS
Genomic Map