Assay Details
| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091058.1 | - | 1 - 2 | 149 | 55 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/arginine repetitive matrix protein 5-like | LOC105378193 | -20.3kb | Anti-sense | Chr. 13 - 112013691 - 112036593 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100506016 | LOC100506016 | 59.5kb | Anti-sense | Chr. 13 - 112155157 - 112160710 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00403
Genomic Map