Assay Details
| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091058.1 | - | 4 - 5 | 406 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SRY-box 1 | SOX1 | -36.2kb | Sense | Chr. 13 - 112067599 - 112071706 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100506016 | LOC100506016 | 44.6kb | Anti-sense | Chr. 13 - 112155157 - 112160710 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00403
Genomic Map