Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZNFX1 antisense RNA 1 | ZFAS1 |
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1 |
-103.1kb | Anti-sense | Chr. 20 - 49278178 - 49289260 on NCBI Build GRCh38 |
| Downstream | prostaglandin I2 (prostacyclin) synthase | PTGIS |
CYP8, CYP8A1, PGIS, PTGI |
108.7kb | Sense | Chr. 20 - 49503874 - 49568170 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map