Assay Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018266.2 | NP_060736.1 | 9 | 116 | ||
| NR_073506.1 | 10 | 116 | ||||
| GenBank mRNA | AK092064.1 | - | 1 | 953 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 31 | ARHGAP31 |
AOS1, CDGAP |
-9.1kb | Sense | Chr. 3 - 119294289 - 119419476 on NCBI Build GRCh38 |
| Downstream | protein O-glucosyltransferase 1 | POGLUT1 |
C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46 |
38.4kb | Sense | Chr. 3 - 119468938 - 119494708 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

TMEM39A
Genomic Map