Assay Details
| Gene Symbol: LOC105378499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092127.1 | - | 1 | 543 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 10 open reading frame 82 | C10orf82 | -160.0 | Anti-sense | Chr. 10 - 116662211 - 116670305 on NCBI Build GRCh38 | |
| Downstream | enolase family member 4 | ENO4 | C10orf134 | 176.5kb | Sense | Chr. 10 - 116848230 - 116911803 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105378499
Genomic Map