Assay Details
| Gene Symbol: RAB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002867.3 | NP_002858.2 | 5 | 2755 | 92 | |
| XM_017001958.1 | XP_016857447.1 | 5 | 2904 | 92 | ||
| GenBank mRNA | AK092167.1 | - | 1 | 1546 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nardilysin convertase | NRDC |
NRD1, hNRD1, hNRD2 |
-38.8kb | Sense | Chr. 1 - 51789192 - 51878937 on NCBI Build GRCh38 |
| Downstream | thioredoxin domain containing 12 | TXNDC12 |
AG1, AGR1, ERP16, ERP18, ERP19, PDIA16, TLP19, hAG-1, hTLP19 |
100.5kb | Sense | Chr. 1 - 52020131 - 52056171 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

RAB3B
Genomic Map