Assay Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304777.1 | NP_001291706.1 | 7 | 3984 | 105 | |
| NM_001304779.1 | NP_001291708.1 | 4 | 3623 | 105 | ||
| NM_016041.4 | NP_057125.2 | 7 | 3987 | 105 | ||
| NR_130905.1 | 6 | 4099 | 105 | |||
| NR_130906.1 | 6 | 3893 | 105 | |||
| GenBank mRNA | AK092218.1 | - | 1 | 1753 | 105 | |
| BM675359.1 | - | 1 | 105 | |||
| CA425310.1 | - | 1 | 105 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371506 | LOC105371506 | -865.0 | Anti-sense | Chr. 17 - 5469092 - 5470360 on NCBI Build GRCh38 | |
| Downstream |
MIS12, kinetochore complex component |
MIS12 |
2510025F08Rik, KNTC2AP, MTW1, hMis12 |
13.1kb | Anti-sense | Chr. 17 - 5486374 - 5490814 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DERL2
Genomic Map