Assay Details
| Gene Symbol: TGM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092657.1 | - | 1 | 1149 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 15 | KIF15 |
HKLP2, KNSL7, NY-BR-62 |
-43.8kb | Sense | Chr. 3 - 44761714 - 44853274 on NCBI Build GRCh38 |
| Downstream | zinc finger DHHC-type containing 3 | ZDHHC3 |
DHHC-3, GODZ, ZNF373 |
664.0 | Anti-sense | Chr. 3 - 44915261 - 44976190 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000471637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000471637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000296125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000422219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000459830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000471637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000471637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000490928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | ENST00000495844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007226.hg.1 | NM_003241.3 | Refseq |

TGM4
Genomic Map