Assay Details
| Gene Symbol: ABHD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093591.1 | - | 1 | 1849 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pleckstrin homology like domain family B member 2 | PHLDB2 |
LL5b, LL5beta |
-2.5kb | Sense | Chr. 3 - 111732480 - 111976517 on NCBI Build GRCh38 |
| Downstream | transgelin 3 | TAGLN3 |
NP22, NP24, NP25 |
17.4kb | Sense | Chr. 3 - 111998739 - 112013888 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABHD10
Genomic Map