Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928441 | LOC101928441 | -781.1kb | Anti-sense | Chr. 12 - 23175636 - 23191587 on NCBI Build GRCh38 | |
| Downstream | microRNA 920 | MIR920 |
MIRN920, hsa-mir-920 |
237.6kb | Anti-sense | Chr. 12 - 24212421 - 24212495 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map