Assay Details
| Gene Symbol: ASCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093971.1 | - | 1 | 1248 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2 |
SPOCK2 | testican-2 | -7.5kb | Sense | Chr. 10 - 72059034 - 72089032 on NCBI Build GRCh38 |
| Downstream | anaphase promoting complex subunit 16 | ANAPC16 |
APC16, C10orf104, CENP-27, MSAG, bA570G20.3 |
117.2kb | Anti-sense | Chr. 10 - 72216000 - 72235860 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ASCC1
Genomic Map