Assay Details
| Gene Symbol: LOC105369662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094655.1 | - | 1 | 2047 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BLOC-1 related complex subunit 5 | BORCS5 |
LOH12CR1, LOH1CR12 |
-12.3kb | Sense | Chr. 12 - 12357078 - 12469562 on NCBI Build GRCh38 |
| Downstream | cAMP responsive element binding protein like 2 | CREBL2 | 128.0kb | Sense | Chr. 12 - 12611833 - 12645108 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105369662
Genomic Map