Assay Details
| Gene Symbol: LOC105378967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095157.1 | - | 1 | 2247 | 84 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB310312.1 | - | 1 | Chr. 5 - 54645457 - 54645987 [-] on Build GRCh38 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 18 | SNX18 |
SH3PX2, SH3PXD3B, SNAG1 |
-97.0kb | Sense | Chr. 5 - 54517759 - 54619249 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102467080 | LOC102467080 | 14.2kb | Anti-sense | Chr. 5 - 54660158 - 54744258 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105378967
Genomic Map