Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102546226 | LOC102546226 | -96.9kb | Sense | Chr. 5 - 88408982 - 88439090 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 461 | LINC00461 |
EyeLinc1, VISC, Visc-1a, Visc-1b, Visc-2 |
3.5kb | Sense | Chr. 5 - 88540779 - 88684803 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map