| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000554901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000556607.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
ENST00000557566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009787.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172263.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172263.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172263.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172263.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172263.hg.1 |
TC1400009787.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400172264.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172264.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172264.hg.1 |
ENST00000216484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172264.hg.1 |
NM_004863.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172264.hg.1 |
TC1400009787.hg.1 |
Clariom D TC |