Assay Details
| Gene Symbol: GALNT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006718225.2 | XP_006718288.1 | 7 | 2596 | 108 | |
| GenBank mRNA | CR627308.1 | - | 1 | 6743 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kinase 2 alpha 3 | CSNK2A3 | CSNK2A1P | -8.7kb | Sense | Chr. 11 - 11351942 - 11353357 on NCBI Build GRCh38 |
| Downstream | microRNA 4299 | MIR4299 | 285.5kb | Sense | Chr. 11 - 11656651 - 11656722 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GALNT18
Genomic Map