Assay Details
| Gene Symbol: NCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190796.2 | NP_001177725.1 | 3 | 4102 | 68 | |
| NM_001291999.1 | NP_001278928.1 | 4 | 4372 | 68 | ||
| NM_006153.5 | NP_006144.1 | 4 | 4338 | 68 | ||
| GenBank mRNA | AK095534.1 | - | 1 | 2248 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NCK1 antisense RNA 1 (head to head) | NCK1-AS1 | SLC35G2-AS1 | -87.2kb | Anti-sense | Chr. 3 - 136841976 - 136862063 on NCBI Build GRCh38 |
| Downstream | interleukin 20 receptor subunit beta | IL20RB |
DIRS1, FNDC6, IL-20R2 |
6.3kb | Sense | Chr. 3 - 136957865 - 137011085 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NCK1
Genomic Map