Assay Details
| Gene Symbol: MOCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004531.4 | NP_004522.1 | 7 | 3402 | 79 | |
| NM_176806.3 | NP_789776.1 | 7 | 2941 | 79 | ||
| GenBank mRNA | AK095832.1 | - | 1 | 1651 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin subunit alpha 2 | ITGA2 |
BR, CD49B, GPIa, HPA-5, VLA-2, VLAA2 |
-1.0kb | Anti-sense | Chr. 5 - 52989326 - 53094779 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC257396 | LOC257396 | 11.7kb | Anti-sense | Chr. 5 - 53109842 - 53115126 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MOCS2
Genomic Map