Assay Details
| Gene Symbol: MCPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534755.2 | XP_011533057.1 | 14 | 3269 | 81 | |
| XM_011534760.2 | XP_011533062.1 | 10 | 2845 | 81 | ||
| XM_017013829.1 | XP_016869318.1 | 15 | 3411 | 81 | ||
| XM_017013831.1 | XP_016869320.1 | 12 | 3067 | 81 | ||
| XM_017013832.1 | XP_016869321.1 | 11 | 2989 | 81 | ||
| GenBank mRNA | AK095865.1 | - | 1 | 2155 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8055 | MIR8055 | hsa-mir-8055 | -2.3kb | Anti-sense | Chr. 8 - 6622124 - 6622220 on NCBI Build GRCh38 |
| Downstream | 1-acylglycerol-3-phosphate O-acyltransferase 5 | AGPAT5 |
1AGPAT5, LPAATE |
81.4kb | Sense | Chr. 8 - 6708357 - 6761503 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MCPH1
Genomic Map