Assay Details
| Gene Symbol: CSNK1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096950.1 | - | 1 - 2 | 468 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 16 member 3 | SLC16A3 |
MCT 3, MCT 4, MCT-3, MCT-4, MCT3, MCT4 |
-3.2kb | Anti-sense | Chr. 17 - 82228406 - 82239499 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929511 | LOC101929511 | 44.6kb | Sense | Chr. 17 - 82290046 - 82292814 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CSNK1D
Genomic Map