Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cancer susceptibility candidate 15 (non-protein coding) | CASC15 | LINC00340 | Sense | Chr. 6 - 21666444 - 22194400 on NCBI Build GRCh38 | |
| Downstream | neuroblastoma associated transcript 1 | NBAT1 |
CASC14, NBAT-1 |
167.3kb | Anti-sense | Chr. 6 - 22134602 - 22147193 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map