Assay Details
| Gene Symbol: LOC105375077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035191.1 | - | 3 | 1519 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
SPT3 homolog, SAGA and STAGA complex component |
SUPT3H |
SPT3, SPT3L |
-193.7kb | Sense | Chr. 6 - 44808844 - 45378051 on NCBI Build GRCh38 |
| Downstream | chloride intracellular channel 5 | CLIC5 |
DFNB102, DFNB103, MST130, MSTP130 |
325.7kb | Sense | Chr. 6 - 45898451 - 46129809 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105375077
Genomic Map