Assay Details
| Gene Symbol: LOC101928933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC039325.1 | - | 1 | 652 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | torsin 1A interacting protein 1 | TOR1AIP1 |
LAP1, LAP1B |
-33.1kb | Anti-sense | Chr. 1 - 179882042 - 179920077 on NCBI Build GRCh38 |
| Downstream | centrosomal protein 350 | CEP350 |
CAP350, GM133 |
333.0 | Anti-sense | Chr. 1 - 179954773 - 180114880 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928933
Genomic Map