Assay Details
| Gene Symbol: LOC101929133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040618.1 | - | 1 - 2 | 232 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD-box helicase 6 | DDX6 |
HLR2, P54, RCK |
-94.9kb | Sense | Chr. 11 - 118747763 - 118791263 on NCBI Build GRCh38 |
| Downstream | B-cell CLL/lymphoma 9-like | BCL9L |
BCL9-2, DLNB11 |
8.4kb | Sense | Chr. 11 - 118896140 - 118926072 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929133
Genomic Map