Assay Details
| Gene Symbol: LINC01209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110819.1 | 1 - 2 | 828 | 83 | ||
| GenBank mRNA | BC041880.1 | - | 1 - 2 | 829 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1208 | LINC01208 | -178.6kb | Sense | Chr. 3 - 176604148 - 176635532 on NCBI Build GRCh38 | |
| Downstream | transducin (beta)-like 1 X-linked receptor 1 | TBL1XR1 |
C21, DC42, IRA1, MRD41, TBLR1 |
203.8kb | Sense | Chr. 3 - 177019355 - 177197357 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01209
Genomic Map