Assay Details
| Gene Symbol: LOC105378604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR936739.1 | - | 1 | 4149 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4251 | MIR4251 | -178.6kb | Anti-sense | Chr. 1 - 3127975 - 3128035 on NCBI Build GRCh38 | |
| Downstream | Rho guanine nucleotide exchange factor 16 | ARHGEF16 |
GEF16, NBR |
142.9kb | Anti-sense | Chr. 1 - 3454583 - 3481115 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105378604
Genomic Map