Assay Details
| Gene Symbol: LOC101929057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111915.1 | 2 - 3 | 405 | 74 | ||
| GenBank mRNA | CR936797.1 | - | 2 - 3 | 405 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LYR motif containing 2 | LYRM2 | DJ122O8.2 | -36.6kb | Anti-sense | Chr. 6 - 89632224 - 89638755 on NCBI Build GRCh38 |
| Downstream | caspase 8 associated protein 2 | CASP8AP2 |
CED-4, FLASH, RIP25 |
145.8kb | Sense | Chr. 6 - 89829900 - 89874436 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929057
Genomic Map