Assay Details
| Gene Symbol: ATAD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011532924.2 | XP_011531226.1 | 24 | 5462 | 75 | |
| GenBank mRNA | AK027272.2 | - | 1 | 1557 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kelch like family member 29 | KLHL29 | KBTBD9 | -63.0kb | Anti-sense | Chr. 2 - 23385419 - 23708613 on NCBI Build GRCh38 |
| Downstream | UBX domain protein 2A | UBXN2A | UBXD4 | 153.7kb | Anti-sense | Chr. 2 - 23927201 - 24001000 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ATAD2B
Genomic Map