Assay Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129844.1 | - | 1 | 1447 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centrosomal protein 85 like | CEP85L |
C6orf204, NY-BR-15, bA57K17.2 |
Anti-sense | Chr. 6 - 118460772 - 118710101 on NCBI Build GRCh38 | |
| Downstream | selenoprotein K pseudogene | LOC100287632 | 220.6kb | Sense | Chr. 6 - 118782708 - 118783420 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PLN
Genomic Map