Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 88 | WDR88 | PQWD | -20.3kb | Sense | Chr. 19 - 33132092 - 33175799 on NCBI Build GRCh38 |
| Downstream | solute carrier family 7 member 10 | SLC7A10 |
ASC1, HASC-1, asc-1 |
9.2kb | Anti-sense | Chr. 19 - 33208664 - 33225850 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map