Assay Details
Gene Symbol: LINC00943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY070435.1 | - | 1165 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100996671 | LOC100996671 | -54.5kb | Sense | Chr. 12 - 126652946 - 126690318 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC440117 | LOC440117 | 124.2kb | Sense | Chr. 12 - 126869494 - 126874690 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000345111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000535544.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000537374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000539315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000540250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000541359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | ENST00000616373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009332.hg.1 | NR_038256.1 | Refseq |
