Assay Details
| Gene Symbol: LOC101929977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC032907.2 | - | 1 | 648 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carboxymethylenebutenolidase homolog | CMBL | JS-1 | -44.7kb | Sense | Chr. 5 - 10277595 - 10308056 on NCBI Build GRCh38 |
| Downstream | membrane associated ring-CH-type finger 6 | MARCH6 |
DOA10, MARCH-VI, RNF176, TEB4 |
38.0 | Anti-sense | Chr. 5 - 10353639 - 10440388 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929977
Genomic Map