Assay Details
| Gene Symbol: LOC101927402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136167.1 | 1 | 1209 | 76 | ||
| GenBank mRNA | BC035180.2 | - | 1 | 1147 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | galectin like | LGALSL |
GRP, HSPC159 |
-60.8kb | Anti-sense | Chr. 2 - 64454193 - 64461383 on NCBI Build GRCh38 |
| Downstream | aftiphilin | AFTPH | Nbla10388 | 212.0 | Anti-sense | Chr. 2 - 64524305 - 64593005 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927402
Genomic Map