Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ENTPD1 antisense RNA 1 | ENTPD1-AS1 | -17.9kb | Sense | Chr. 10 - 95753206 - 96090235 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 518A | ZNF518A | ZNF518 | 18.9kb | Anti-sense | Chr. 10 - 96129220 - 96165512 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map